Phenylpyruvic acid sodium salt monohydrate 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
2-Oxo-3-phenylpropionic acid sodium salt monohydrate, Sodium phenylpyruvate monohydrate, Sodium 2-oxo-3-phenylpropanoate monohydrate
A14027.03
A14027.06
A14027.14
A14027.18
A14027.03EA
22.7
EUR
A14027.03
A14027.06
A14027.14
A14027.18
Phenylpyruvic acid sodium salt monohydrate 98%
Phenylpyruvic acid sodium salt monohydrate
Purity: 98%
CAS No.: 122049-54-1
Molecular Formula: C9H7NaO3.H2O
Formula Weight: 204.16 (186.14anhy)
Storage Temperature: 2°C to 8°C
Melting Point:
Formel:
C₉H₉NaO₄ Molecular Weight: 204.157 g/mol |
MDL:
MFCD00149109 CAS-Nummer: 122049-54-1 EINECS: 204-053-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...