4-tert-Butylthiazol-2-ylamine 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Amino-4-(tert-butyl)-1,3-thiazole

A14115.03 A14115.06 A14115.14
A14115.03EA 20.6 EUR
A14115.03 A14115.06 A14115.14
4-tert-Butylthiazol-2-ylamine 98%
4-tert-Butylthiazol-2-ylamine

Purity: 98% CAS No.: 74370-93-7 Molecular Formula: C7H12N2S Formula Weight: 156.26 Melting Point: 98-103° Beilstein Reference: 114270 MDL No.: MFCD00051644

Formel: C₇H₁₂N₂S
Molecular Weight: 156,25 g/mol
Schmelzpunkt: 98…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051644
CAS-Nummer: 74370-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR