6-Aza-2-thiothymine 98%
Lieferant: Thermo Scientific
Synonyme:
6-Methyl-3-thioxo-3,4-dihydro-1,2,4-triazin-5(2H)-one
A14167.06
A14167.14
A14167.22
A14167.06EA
66.6
EUR
A14167.06
A14167.14
A14167.22
6-Aza-2-thiothymine 98%
6-Aza-2-thiothymine
Purity: 98%
CAS No.: 615-76-9
Molecular Formula: C4H5N3OS
Formula Weight: 143.17
Melting Point: 218-221°
Beilstein Reference: 4861
MDL No.: MFCD00006458
Formel:
C₄H₅N₃OS Molecular Weight: 143,17 g/mol Schmelzpunkt: 218…221 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006458 CAS-Nummer: 615-76-9 EINECS: 210-445-4 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...