L-Serine ethyl ester hydrochloride 99%

Lieferant: Thermo Fisher Scientific

Synonyme: H-Ser-OEt.HCl, H-Ser-OEt·HCl

A14174.06 A14174.14
A14174.06EA 54.4 EUR
A14174.06 A14174.14
L-Serine ethyl ester hydrochloride 99%
L-Serine ethyl ester hydrochloride

Purity: 99% CAS No.: 26348-61-8 Molecular Formula: C5H11NO3.HCl Formula Weight: 169.61 Storage Temperature: -30°C to -10°C Melting Point: 130-132° Beilstein Reference: 3562346 Optical Rotation: -4.4° (c=4 in water) MDL No.: MFCD00012594

Formel: C₅H₁₂ClNO₃
Molecular Weight: 169.605 g/mol
Schmelzpunkt: 130…132 °C
MDL: MFCD00012594
CAS-Nummer: 26348-61-8
EINECS: 247-624-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR