Bis(3-methyl-1-phenyl-5-pyrazolone) ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,3-Dimethyl-1,1-diphenyl-1H,1H-[4,4-bipyrazole]-5,5(4H,4H)-dione, 3,3'-Dimethyl-1,1'-diphenyl-4,4'-bi(5-pyrazolone), Bispyrazolone
A14308.06
A14308.14
A14308.06EA
44
EUR
A14308.06
A14308.14
Bis(3-methyl-1-phenyl-5-pyrazolone) ≥98%
Bis(3-methyl-1-phenyl-5-pyrazolone)
Purity: 98+%
CAS No.: 7477-67-0
Molecular Formula: C20H18N4O2
Formula Weight: 346.39
Melting Point:
Formel:
C₂₀H₁₈N₄O₂ Molecular Weight: 346.39 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020765 CAS-Nummer: 7477-67-0 EINECS: 231-282-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...