5,6-Dichloro-2-mercaptobenzimidazole 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5,6-Dichloro-1H-benzimidazole-2-thiol

A14309.03 A14309.06
A14309.03EA 57.1 EUR
A14309.03 A14309.06
5,6-Dichloro-2-mercaptobenzimidazole 98%
5,6-Dichloro-2-mercaptobenzimidazole

Purity: 98% CAS No.: 19462-98-7 Molecular Formula: C7H4Cl2N2S Formula Weight: 219.09 Odour: Odourless Melting Point: ca 350° dec. MDL No.: MFCD00052398

Formel: C₇H₄Cl₂N₂S
Molecular Weight: 219,09 g/mol
Schmelzpunkt: ∼ 350 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00052398
CAS-Nummer: 19462-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR