Lawesson-Reagenz 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,4-Bis(4-methoxyphenyl)-2,4-dithioxo-1,3,2,4- dithiadiphosphetane, 2,4-Bis(4-methoxyphenyl)-1,3-dithia-2,4-diphosphetane 2,4-Disulphide
A14530.06
A14530.14
A14530.22
A14530.36
A14530.06EA
20.6
EUR
A14530.06
A14530.14
A14530.22
A14530.36
Lawesson-Reagenz 97%
Lawesson's reagent
Purity: 97%
CAS No.: 19172-47-5
Molecular Formula: C14H14O2P2S4
Formula Weight: 404.47
Melting Point: 221-225°
Beilstein Reference: 1024888
UN No.: UN3134
MDL No.: MFCD00005171
Formel:
C₁₄H₁₄O₂P₂S₄ Molecular Weight: 404,45 g/mol Schmelzpunkt: 221…225 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005171 CAS-Nummer: 19172-47-5 EINECS: 242-855-4 ADR: 4.3,III Merck Index: 14,5391 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...