3-Octen-2-on 97%

Lieferant: Thermo Fisher Scientific

A14652.09 A14652.18 A14652.30
A14652.09EA 36.4 EUR
A14652.09 A14652.18 A14652.30
3-Octen-2-on 97%
3-Octen-2-on

Purity: 97% CAS No.: 1669-44-9 Molecular Formula: C8H14O Formula Weight: 126.20 Flash Point: 54°(129°F) Refractive Index: 1.4480 Odour: Pleasant Boiling Point: 60°/16mm Beilstein Reference: 1742230 UN No.: UN1224 MDL No.: MFCD00015565

Formel: C₈H₁₄O
Molecular Weight: 126,20 g/mol
MDL: MFCD00015565
CAS-Nummer: 1669-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR