(S)-Methyl-2-amino-4-methylpentanoate hydrochloride 99%

Lieferant: Thermo Scientific

Synonyme: H-Leu-OMe·HCl, H-Leu-OMe.HCl, L-Leucine methyl ester hydrochloride

A14897.14 A14897.22 A14897.36
A14897.14EA 57.3 EUR
A14897.14 A14897.22 A14897.36
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride 99%
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride

Purity: 99% CAS No.: 7517-19-3 Molecular Formula: C7H15NO2.HCl Formula Weight: 181.66 Melting Point: 149-151° Beilstein Reference: 3595133 Optical Rotation: +13° (c=2 in water) MDL No.: MFCD00012494

Formel: C₇H₁₅NO₂·HCl
Molecular Weight: 181,7 g/mol
Schmelzpunkt: 149…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012494
CAS-Nummer: 7517-19-3
EINECS: 231-375-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR