2,2'-(2,5-Thiendiyl)bis[5-(2-methyl-2-propanyl)-1,3-benzoxazol] 99%

Lieferant: Thermo Fisher Scientific

Synonyme: 2,5-Bis(5-(tert-butyl)benzo[d]oxazol-2-yl)thiophene

A14928.06 A14928.14 A14928.22
A14928.06EA 47.9 EUR
A14928.06 A14928.14 A14928.22
2,2'-(2,5-Thiendiyl)bis[5-(2-methyl-2-propanyl)-1,3-benzoxazol] 99%
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene

Purity: 99% CAS No.: 7128-64-5 Molecular Formula: C26H26N2O2S Formula Weight: 430.57 Melting Point: 200-202° Beilstein Reference: 631803 MDL No.: MFCD00005774

Formel: C₂₆H₂₆N₂O₂S
Molecular Weight: 430.57 g/mol
Schmelzpunkt: 200…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005774
CAS-Nummer: 7128-64-5
EINECS: 230-426-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR