Cholesteryl-acetat ≥97%
Lieferant: Thermo Scientific
Synonyme:
(3S,8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl acetate, Cholesteryl Acetate, Acetyl Cholesterol, Acetic Acid Cholesterol Ester
A15052.14
A15052.22
A15052.36
A15052.14EA
97.2
EUR
A15052.14
A15052.22
A15052.36
Cholesteryl-acetat ≥97%
Cholesterol acetate
Purity: 97+%
CAS No.: 604-35-3
Molecular Formula: C29H48O2
Formula Weight: 428.70
Melting Point: 112-114°
Beilstein Reference: 2064235
Optical Rotation: -43° (c=5 in chloroform)
MDL No.: MFCD00003636
Formel:
C₂₉H₄₈O₂ Molecular Weight: 428,70 g/mol Schmelzpunkt: 112…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003636 CAS-Nummer: 604-35-3 EINECS: 210-066-4 Merck Index: 14,2201 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...