Ethylenediaminetetraacetic acid diammonium salt hydrate

Lieferant: Thermo Fisher Scientific

Synonyme: EDTA diammonium salt hydrate

A15286.14 A15286.22 A15286.36
A15286.14EA 23 EUR
A15286.14 A15286.22 A15286.36
Ethylenediaminetetraacetic acid diammonium salt hydrate
Ethylenediaminetetraacetic acid diammonium salt hydrate

CAS No.: 20824-56-0 Molecular Formula: (NH4O2CCH2)2NCH2CH2N(CH2CO2H)2.xH2O Formula Weight: 326.31(anhy) Beilstein Reference: 7785165 MDL No.: MFCD00150463

Formel: C₁₀H₂₂N₄O₈·xH₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00150463
CAS-Nummer: 20824-56-0
EINECS: 244-063-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR