6-Bromo-2-oxo-2H-chromene-3-carboxylic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6-Bromocoumarin-3-carboxylic acid, UBP608
A15354.06
A15354.14
A15354.06EA
91.6
EUR
A15354.06
A15354.14
6-Bromo-2-oxo-2H-chromene-3-carboxylic acid 97%
6-Bromo-2-oxo-2H-chromene-3-carboxylic acid
Purity: 97%
CAS No.: 2199-87-3
Molecular Formula: C10H5BrO4
Formula Weight: 269.05
Melting Point: 194-196°
Beilstein Reference: 13960
MDL No.: MFCD00047640
Formel:
C₁₀H₅BrO₄ Molecular Weight: 269.06 g/mol Schmelzpunkt: 194…196 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047640 CAS-Nummer: 2199-87-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...