Terephthalaldehyde mono(diethyl acetal) 98% stabilisiert

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(Diethoxymethyl)benzaldehyde

A15413.06 A15413.14 A15413.22 A15413.36
A15413.06EA 44.9 EUR
A15413.06 A15413.14 A15413.22 A15413.36
Terephthalaldehyde mono(diethyl acetal) 98% stabilisiert
Terephthalaldehyde mono(diethyl acetal)

Purity: 98% CAS No.: 81172-89-6 Molecular Formula: C12H16O3 Formula Weight: 208.26 Flash Point: 110°(230°F) Refractive Index: 1.5070 Boiling Point: 89-93°/7mm Beilstein Reference: 4293872 MDL No.: MFCD00010217

Formel: C₁₂H₁₆O₃
Molecular Weight: 208,26 g/mol
Siedepunkt: 89…93 °C (7 mmHg)
Dichte: 1,047
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010217
CAS-Nummer: 81172-89-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR