3-(1-Naphthyl)acrylic acid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(Naphthalen-1-yl)acrylic acid

A15422.09 A15422.18 A15422.30
A15422.09EA 64.8 EUR
A15422.09 A15422.18 A15422.30
3-(1-Naphthyl)acrylic acid 98%
3-(1-Naphthyl)acrylic acid

Purity: 98% CAS No.: 13026-12-5 Molecular Formula: C13H10O2 Formula Weight: 198.22 Melting Point: 210-212° Beilstein Reference: 1868617 MDL No.: MFCD00014317

Formel: C₁₃H₁₀O₂
Molecular Weight: 198,22 g/mol
Schmelzpunkt: 210…212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014317
CAS-Nummer: 13026-12-5
EINECS: 235-887-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR