1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose ≥98%
Lieferant: Thermo Scientific
Achtung
Synonyme: Diacetone-D-glucose, (-)-1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose, (3aR,5S,6S,6aR)-5-((R)-2,2-Dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol, D-Glucose diacetonide
A15568.14 A15568.22 A15568.36
A15568.14EA 43.9 EUR
A15568.14 A15568.22 A15568.36
1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose ≥98%
1,2:5,6-Di-O-isopropylidene-alpha-D-glucofuranose
Purity: 98+% CAS No.: 582-52-5 Molecular Formula: C12H20O6 Formula Weight: 260.29 Melting Point: 107-110° Beilstein Reference: 84386 Optical Rotation: -11° (c=5 in ethanol) MDL No.: MFCD00005544
Formel: C₁₂H₂₀O₆
Molecular Weight: 260,29 g/mol
Schmelzpunkt: 107…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005544
CAS-Nummer: 582-52-5
EINECS: 209-486-0
Merck Index: 14,2965