meso-Erythritol 99%

Lieferant: Thermo Scientific

Synonyme: meso-Erythritol, (2R,3S)-rel-Butane-1,2,3,4-tetraol, i-Erythritol

A15813.14 A15813.22
A15813.14EA 60.9 EUR
A15813.14 A15813.22
meso-Erythritol 99%
meso-Erythritol

Purity: 99% CAS No.: 149-32-6 Molecular Formula: C4H10O4 Formula Weight: 122.12 Melting Point: 119-124° Boiling Point: 329-331° Beilstein Reference: 1719753 MDL No.: MFCD00004710

Formel: C₄H₁₀O₄
Molecular Weight: 122,12 g/mol
Siedepunkt: 329…331 °C
Schmelzpunkt: 119…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004710
CAS-Nummer: 149-32-6
EINECS: 205-737-3
Merck Index: 14,3675

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR