(1S,2R)-(-)-10,2-Camphorsultam 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: (-)-10,2-Camphorsultam, (2R)-(-)-Bornane-10,2-sultam, (1S,5R)-10,10-Dimethyl-3-thia-4-azatricyclo[5.2.1.0(1,5)]decane 3,3-dioxide, (1S)-(−)-2,10-Camphorsultam, [3aS-(3aalpha,6alpha,7abeta)]-Hexahydro-8,8-dimethyl-2,2-dioxide-3H-3a,6-methano-2,1-benzisothiazole, (3aS,6R,7aR)-8,8-Dimethylhexahydro-1H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide, (3aS,6S,7aS)-8,8-Dimethylhexahydro-1H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide, (1S,2R,4R)-2,10-camphorsultam, (-)-exo-10,2-Bornanesultam, (2R)-Bornane-10,2-sultam

A15897.06 A15897.14
A15897.06EA 142 EUR
A15897.06 A15897.14
(1S,2R)-(-)-10,2-Camphorsultam 99%
(1S,2R)-(-)-10,2-Camphorsultam

Purity: 99% CAS No.: 94594-90-8 Molecular Formula: C10H17NO2S Formula Weight: 215.32 Melting Point: 181-186° Beilstein Reference: 83811 Optical Rotation: -33° (c=1 in ethanol) MDL No.: MFCD00066271

Formel: C₁₀H₁₇NO₂S
Molecular Weight: 215,32 g/mol
Schmelzpunkt: 181…186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066271
CAS-Nummer: 94594-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR