Boc-L-phenylalanin 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Boc-L-phenylalanine, BOC-L-Phenylalanine, N-α-t-Boc-L-phenylalanine, Boc-Phe-OH
A16017.06
A16017.14
A16017.06EA
34.5
EUR
A16017.06
A16017.14
Boc-L-phenylalanin 99%
Boc-L-phenylalanin
Purity: 99%
CAS No.: 13734-34-4
Molecular Formula: C14H19NO4
Formula Weight: 265.31
Storage Temperature: 2°C to 8°C
Melting Point: 84-88°
Beilstein Reference: 2219729
Optical Rotation: +25° (c=1 in ethanol)
MDL No.: MFCD00002663
Formel:
C₁₄H₁₉NO₄ Molecular Weight: 265.31 g/mol Schmelzpunkt: 84…88 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00002663 CAS-Nummer: 13734-34-4 EINECS: 237-305-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...