N-ɑ-Boc-L-asparagine ≥98%
Lieferant: Thermo Scientific
Synonyme:
Boc-L-Asparagine, Boc-Asn-OH
A16019.06
A16019.14
A16019.06EA
23.1
EUR
A16019.06
A16019.14
N-ɑ-Boc-L-asparagine ≥98%
N-alpha-Boc-L-asparagine
Purity: 98+%
CAS No.: 7536-55-2
Molecular Formula: C9H16N2O5
Formula Weight: 232.24
Storage Temperature: 2°C to 8°C
Melting Point: ca 170° dec.
Beilstein Reference: 1977963
Optical Rotation: -7.3° (c=2 in DMF)
MDL No.: MFCD00038152
Formel:
C₉H₁₆N₂O₅ Molecular Weight: 232,24 g/mol Schmelzpunkt: ∼ 170 °C (Zersetzung) Lagertemperatur: Kühlschrank |
MDL:
MFCD00038152 CAS-Nummer: 7536-55-2 EINECS: 231-405-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...