N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-L-isoleucin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Boc-L-isoleucine, Boc-Ile-OH

A16021.06 A16021.14
A16021.06EA 34.8 EUR
A16021.06 A16021.14
N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-L-isoleucin ≥98%
Boc-L-isoleucine

Purity: 98+% CAS No.: 13139-16-7 Molecular Formula: C11H21NO4 Formula Weight: 231.06 Storage Temperature: 2°C to 8°C Melting Point: 67-70° Beilstein Reference: 1711700 MDL No.: MFCD00038324

Formel: C₁₁H₂₁NO₄
Schmelzpunkt: 67…70 °C
MDL: MFCD00038324
CAS-Nummer: 13139-16-7
EINECS: 236-074-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR