Dithiobiuret 97%

Lieferant: Thermo Fisher Scientific
Gefahr

A16175.06 A16175.14 A16175.22
A16175.06EA 71.3 EUR
A16175.06 A16175.14 A16175.22
Dithiobiuret 97%
Dithiobiuret

Purity: 97% CAS No.: 541-53-7 Molecular Formula: C2H5N3S2 Formula Weight: 135.21 Odour: Odourless Melting Point: ca 183° dec. Beilstein Reference: 1756621 UN No.: UN2811 MDL No.: MFCD00037832

Formel: C₂H₅N₃S₂
Molecular Weight: 135,21 g/mol
Schmelzpunkt: ∼ 183 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00037832
CAS-Nummer: 541-53-7
EINECS: 208-784-8
ADR: 6.1,I
Merck Index: 14,3371

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR