Diiodofluorescein
Lieferant: Thermo Fisher Scientific
A16554.09
A16554.18
A16554.09EA
34.7
EUR
A16554.09
A16554.18
Diiodofluorescein
Diiodofluorescein
CAS No.: 31395-16-1
Molecular Formula: C20H10I2O5
Formula Weight: 584.11
Melting Point: 239-241° dec.
MDL No.: MFCD00036461
Formel:
C₂₀H₁₀I₂O₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036461 CAS-Nummer: 31395-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...