D-Pantothenic acid calcium salt hydrate 98%
Lieferant: Thermo Scientific
Synonyme:
Calcium D-pantothenate hydrate
A16609.22
A16609.36
A16609.22EA
52.1
EUR
A16609.22
A16609.36
D-Pantothenic acid calcium salt hydrate 98%
D-Pantothenic acid calcium salt hydrate
Purity: 98%
CAS No.: 331748-07-3
Molecular Formula: C18H32CaN2O10.xH2O
Formula Weight: 476.54(anhy)
Melting Point: ca 138° dec.
Beilstein Reference: 3769272
Optical Rotation: +27° (c=5 in water)
MDL No.: MFCD00002766
Formel:
C₁₈H₃₂CaN₂O₁₀·xH₂O Molecular Weight: 476,54 g/mol (anhy) Schmelzpunkt: ∼ 138 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002766 CAS-Nummer: 331748-07-3 EINECS: 205-278-9 Merck Index: 14,7015 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...