4-(Dimethylamino)butanoic acid hydrochloride 98%

Lieferant: Thermo Scientific
Achtung

A16610.06 A16610.14
A16610.06EA 48.7 EUR
A16610.06 A16610.14
4-(Dimethylamino)butanoic acid hydrochloride 98%
4-(Dimethylamino)butanoic acid hydrochloride

Purity: 98% CAS No.: 69954-66-1 Molecular Formula: HO2C(CH2)3N(CH3)2.HCl Formula Weight: 167.64 Melting Point: 153-155° MDL No.: MFCD00012613

Formel: C₆H₁₄ClNO₂
Schmelzpunkt: 153…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012613
CAS-Nummer: 69954-66-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR