3-Noradamantanecarboxylic acid 98%

Lieferant: Thermo Scientific
Achtung

A16683.01 A16683.03 A16683.06
A16683.01EA 26.2 EUR
A16683.01 A16683.03 A16683.06
3-Noradamantanecarboxylic acid 98%
3-Noradamantanecarboxylic acid

Purity: 98% CAS No.: 16200-53-6 Molecular Formula: C10H14O2 Formula Weight: 166.22 Melting Point: 105-107° MDL No.: MFCD00213463

Formel: C₁₀H₁₄O₂
Molecular Weight: 166,22 g/mol
Schmelzpunkt: 105…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00213463
CAS-Nummer: 16200-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR