N-[(Benzyloxy)carbonyl]-L-alanin 98%
Lieferant: Thermo Scientific
Synonyme:
N-Cbz-L-alanine, Z-Ala-OH, (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid, N-Benzyloxycarbonyl-L-alanine, ((Benzyloxy)carbonyl)-L-alanine
A16716.06
A16716.14
A16716.22
A16716.06EA
28.8
EUR
A16716.06
A16716.14
A16716.22
N-[(Benzyloxy)carbonyl]-L-alanin 98%
N-Carbobenzyloxy-L-alanine
Purity: 98%
CAS No.: 1142-20-7
Molecular Formula: C11H13NO4
Formula Weight: 223.23
Melting Point: 85-87°
Beilstein Reference: 2056164
Optical Rotation: -14.5° (c=2 in acetic acid)
MDL No.: MFCD00002640
Formel:
C₁₁H₁₃NO₄ Molecular Weight: 223,23 g/mol Schmelzpunkt: 85…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002640 CAS-Nummer: 1142-20-7 EINECS: 214-532-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...