1-Phenylmethansulfonamid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Alpha-toluenesulphonamide
A16740.06
A16740.03
A16740.06EA
154
EUR
A16740.06
A16740.03
1-Phenylmethansulfonamid 98%
Phenylmethanesulphonamide
Purity: 98%
CAS No.: 4563-33-1
Molecular Formula: C7H9NO2S
Formula Weight: 171.22
Melting Point: 101-104°
MDL No.: MFCD00041893
Formel:
C₇H₉NO₂S Molecular Weight: 171,22 g/mol Schmelzpunkt: 101…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041893 CAS-Nummer: 4563-33-1 EINECS: 224-935-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...