Ethyl-2-thiouracil-5-carboxylate 99%

Lieferant: Thermo Scientific

Synonyme: 5-Carbethoxy-2-thiouracil

A17046.06 A17046.14
A17046.06EA 29.6 EUR
A17046.06 A17046.14
Ethyl-2-thiouracil-5-carboxylate 99%
Ethyl-2-thiouracil-5-carboxylate

Purity: 99% CAS No.: 38026-46-9 Molecular Formula: C7H8N2O3S Formula Weight: 200.22 Melting Point: ca 250° dec. Beilstein Reference: 190867 MDL No.: MFCD00006074

Formel: C₇H₈N₂O₃S
Molecular Weight: 200,22 g/mol
Schmelzpunkt: ∼ 250 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006074
CAS-Nummer: 38026-46-9
EINECS: 253-749-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR