N-Carbamoyl-DL-aspartic acid 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-Ureidosuccinic acid, N-Carbamyl-DL-aspartic Acid

A17166.18 A17166.09
A17166.18EA 1157 EUR
A17166.18 A17166.09
N-Carbamoyl-DL-aspartic acid 98%
N-Carbamoyl-DL-aspartic acid

Purity: 98% CAS No.: 923-37-5 Molecular Formula: C5H8N2O5 Formula Weight: 176.13 Melting Point: ca 175° dec. Beilstein Reference: 1726861 MDL No.: MFCD00042822

Formel: C₅H₈N₂O₅
Molecular Weight: 176.13 g/mol
Schmelzpunkt: ∼ 175 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042822
CAS-Nummer: 923-37-5
EINECS: 213-096-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR