(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Cbz-Phe-OH, N-Cbz-L-phenylalanine, N-Carbobenzyloxy-L-phenylalanine, Z-Phe-OH
A17182.06
A17182.14
A17182.06EA
31.4
EUR
A17182.06
A17182.14
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid 99%
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid
Purity: 99%
CAS No.: 1161-13-3
Molecular Formula: C17H17NO4
Formula Weight: 299.33
Melting Point: 86-92°
Beilstein Reference: 2222826
Optical Rotation: +5° (c=5 in acetic acid)
MDL No.: MFCD00020418
Formel:
C₁₇H₁₇NO₄ Molecular Weight: 299.3 g/mol Schmelzpunkt: 86…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020418 CAS-Nummer: 1161-13-3 EINECS: 214-599-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...