Methyl-L-Lysinat-Dihydrochlorid 99%

Lieferant: Thermo Scientific

Synonyme: H-Lys-OMe·2HCl, H-Lys-OMe.2HCl

A18157.14 A18157.22 A18157.36
A18157.14EA 71.1 EUR
A18157.14 A18157.22 A18157.36
Methyl-L-Lysinat-Dihydrochlorid 99%
Methyl-L-Lysinat-Dihydrochlorid

Purity: 99% CAS No.: 26348-70-9 Molecular Formula: C7H16N2O2.2HCl Formula Weight: 233.14 Storage Temperature: 2°C to 8°C Melting Point: ca 200° dec. Beilstein Reference: 5302183 MDL No.: MFCD00039067

Formel: C₇H₁₆N₂O₂
Molecular Weight: 233,13 g/mol
Schmelzpunkt: ∼ 200 °C (Zersetzung)
Lagertemperatur: Kühlschrank
MDL: MFCD00039067
CAS-Nummer: 26348-70-9
EINECS: 247-625-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR