(5-Nitro-2-furyl)methylen-diacetat 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Nitrofurfural Diacetyl Acetal, Nitrofurfural diacetate, 2-(Diacetoxymethyl)-5-nitrofuran, (5-Nitrofuran-2-yl)methylene diacetate, 5-Nitrofurfurylidene Diacetate

A18169.22 A18169.36
A18169.22EA 32.9 EUR
A18169.22 A18169.36
(5-Nitro-2-furyl)methylen-diacetat 97%
(5-Nitro-2-furyl)methylen-diacetat

Purity: 98+% CAS No.: 92-55-7 Molecular Formula: C9H9NO7 Formula Weight: 243.17 Melting Point: 87-92° Beilstein Reference: 255089 MDL No.: MFCD00003244

Formel: C₉H₉NO₇
Molecular Weight: 243,17 g/mol
Schmelzpunkt: 87…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003244
CAS-Nummer: 92-55-7
EINECS: 202-166-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR