Indium(III) phosphate 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A18281.06
A18281.14
A18281.06EA
47.4
EUR
A18281.06
A18281.14
Indium(III) phosphate 98%
Indium(III) phosphate
Purity: 98%
CAS No.: 14693-82-4
Molecular Formula: InPO4
Formula Weight: 209.79
MDL No.: MFCD00016154
Formel:
InPO₄ Dichte: 4,9 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016154 CAS-Nummer: 14693-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...