4-Acetyldiphenyl sulphone 98%

Lieferant: Thermo Scientific

A18376.03 A18376.06 A18376.14
A18376.03EA 84.7 EUR
A18376.03 A18376.06 A18376.14
4-Acetyldiphenyl sulphone 98%
4-Acetyldiphenyl sulphone

Purity: 98% CAS No.: 65085-83-8 Molecular Formula: C14H12O3S Formula Weight: 260.31 Melting Point: 138-140° MDL No.: MFCD00209628

Formel: C₁₄H₁₂O₃S
Molecular Weight: 260,31 g/mol
Schmelzpunkt: 138…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00209628
CAS-Nummer: 65085-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR