Cyclohexylamine hydrobromide 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Cyclohexanamine hydrobromide, Cyclohexylamine HBr
A18439.22
A18439.36
A18439.22EA
34.6
EUR
A18439.22
A18439.36
Cyclohexylamine hydrobromide 98%
Cyclohexylamine hydrobromide
Purity: 98%
CAS No.: 26227-54-3
Molecular Formula: C6H13N.HBr
Formula Weight: 180.09
Melting Point: 197-201°
Beilstein Reference: 3908098
MDL No.: MFCD00034986
Formel:
C₆H₁₃N·HBr Molecular Weight: 180,09 g/mol Schmelzpunkt: 197…201 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00034986 CAS-Nummer: 26227-54-3 EINECS: 247-526-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...