Sodium-DL-2-hydroxybutyrate ≥97%

Lieferant: Thermo Scientific

Synonyme: DL-2-Hydroxybutyric Acid Sodium Salt, Sodium 2-hydroxybutanoate

A18636.03 A18636.06 A18636.14
A18636.03EA 26.8 EUR
A18636.03 A18636.06 A18636.14
Sodium-DL-2-hydroxybutyrate ≥97%
Sodium-DL-2-hydroxybutyrate

Purity: 97+% CAS No.: 5094-24-6 Molecular Formula: C4H7NaO3 Formula Weight: 126.09 Melting Point: 153-157° Beilstein Reference: 4825104 MDL No.: MFCD00004565

Formel: C₄H₈O₃
Molecular Weight: 126,09 g/mol
Schmelzpunkt: 153…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004565
CAS-Nummer: 5094-24-6
EINECS: 225-812-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR