N-[(Benzyloxy)carbonyl]serin  99%

Lieferant: Thermo Scientific

Synonyme: N-Carbobenzyloxy-L-serine, Z-Ser-OH, N-Cbz-L-serine

A19050.06 A19050.14 A19050.22
A19050.06EA 41.5 EUR
A19050.06 A19050.14 A19050.22
N-[(Benzyloxy)carbonyl]serin  99%
N-Cbz-L-serine

Purity: 99% CAS No.: 1145-80-8 Molecular Formula: C11H13NO5 Formula Weight: 239.23 Melting Point: 117-120° Beilstein Reference: 2058314 Optical Rotation: +6° (c=7 in acetic acid) MDL No.: MFCD00002662

Formel: C₁₁H₁₃NO₅
Molecular Weight: 239,2 g/mol
Schmelzpunkt: 117…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002662
CAS-Nummer: 1145-80-8
EINECS: 214-546-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR