Sulphamethazine 99%

Lieferant: Thermo Scientific

Synonyme: 4,6-Dimethylsulphadiazine, Sulphadimidine, 4-Amino-N-(4,6-dimethyl-2-pyrimidyl)benzenesulphonamide

A19276.14 A19276.22
A19276.14EA 28.2 EUR
A19276.14 A19276.22
Sulphamethazine 99%
Sulphamethazine

Purity: 99% CAS No.: 57-68-1 Molecular Formula: C12H14N4O2S Formula Weight: 278.34 Melting Point: 198-201° MDL No.: MFCD00006066

Formel: C₁₂H₁₄N₄O₂S
Molecular Weight: 278,33 g/mol
Schmelzpunkt: 198…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006066
CAS-Nummer: 57-68-1
EINECS: 200-346-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR