Ethyl-trans-4-Decenoat 97%
Lieferant: Thermo Scientific
Synonyme:
trans-Obtusilic Acid Ethyl Ester, Ethyl trans-4-Decenoate, trans-4-Decenoic Acid Ethyl Ester
A19364.09
A19364.18
A19364.09EA
41
EUR
A19364.09
A19364.18
Ethyl-trans-4-Decenoat 97%
Ethyl-trans-4-decenoate
Purity: 97%
CAS No.: 76649-16-6
Molecular Formula: CH3(CH2)4CH=CHCH2CH2CO2CH2CH3
Formula Weight: 198.31
Refractive Index: 1.4370
MDL No.: MFCD00015574
Formel:
C₁₂H₂₂O₂ Molecular Weight: 198,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015574 CAS-Nummer: 76649-16-6 EINECS: 278-509-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...