Barium tartrate 98%

Lieferant: Thermo Scientific
Achtung

A19604.18 A19604.30
A19604.18EA 22.7 EUR
A19604.18 A19604.30
Barium tartrate 98%
Barium tartrate

Purity: 98% CAS No.: 5908-81-6 Molecular Formula: C4H4BaO6 Formula Weight: 285.41 Odour: Odourless Solubility: Very slightly soluble in water Beilstein Reference: 6120238 UN No.: UN1564 MDL No.: MFCD00054437

Formel: C₄H₄BaO₆
Molecular Weight: 285,41 g/mol
Dichte: 2,98
Lagertemperatur: Raumtemperatur
MDL: MFCD00054437
CAS-Nummer: 5908-81-6
EINECS: 227-615-9
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR