3-Methylbenzo[b]thiophene-2-acetic acid 97%
Lieferant: Thermo Scientific
|
Achtung
|
A19620.02
A19620.88
A19620.02EA
118
EUR
A19620.02
A19620.88
3-Methylbenzo[b]thiophene-2-acetic acid 97%
3-Methylbenzo[b]thiophene-2-acetic acid
Purity: 97%
CAS No.: 1505-52-8
Molecular Formula: C11H10O2S
Formula Weight: 206.25
Melting Point: 148-150°
MDL No.: MFCD00052299
Formel:
C₁₁H₁₀O₂S Molecular Weight: 206,25 g/mol Schmelzpunkt: 148…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052299 CAS-Nummer: 1505-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...