Ethyl-4,4-imethyl-3-oxovalerate 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 4,4-dimethyl-3-oxopentanoate, Ethyl Pivaloylacetate, 4,4-Dimethyl-3-oxovaleric Acid Ethyl Ester, Pivaloylacetic Acid Ethyl Ester

A19752.22 A19752.14
A19752.22EA 261 EUR
A19752.22 A19752.14
Ethyl-4,4-imethyl-3-oxovalerate 97%
Ethyl-4,4-imethyl-3-oxovalerate

Purity: 97% CAS No.: 17094-34-7 Molecular Formula: (CH3)3CC(O)CH2CO2CH2CH3 Formula Weight: 172.22 Boiling Point: 88-90°/10mm MDL No.: MFCD00042886

Formel: C₉H₁₆O₃
Molecular Weight: 172,22 g/mol
Siedepunkt: 88…90 °C (10 mmHg)
Dichte: 0,967
Lagertemperatur: Raumtemperatur
MDL: MFCD00042886
CAS-Nummer: 17094-34-7
EINECS: 241-162-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR