Citronellylacetat ≥96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,7-Dimethyloct-6-en-1-yl acetate, 3,7-Dimethyl-6-octen-1-ol Acetate

A19872.14 A19872.22
A19872.14EA 24.6 EUR
A19872.14 A19872.22
Citronellylacetat ≥96%
Citronellyl acetate

Purity: 96% CAS No.: 150-84-5 Molecular Formula: C13H24O2 Formula Weight: 212.33 Flash Point: 98°(208°F) Refractive Index: 1.4434 Boiling Point: 240° MDL No.: MFCD00015039

Formel: C₁₂H₂₂O₂
Molecular Weight: 198,31 g/mol
Siedepunkt: 240 °C
Dichte: 0,890
Flammpunkt: 98 °C (208 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00015039
CAS-Nummer: 150-84-5
EINECS: 205-775-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR