3,4-Dimethyl-1-pentin-3-ol 94%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(1,2-Dimethyl-1-hydroxypropyl)acetylene
A19906.06
A19906.14
A19906.06EA
33.7
EUR
A19906.06
A19906.14
3,4-Dimethyl-1-pentin-3-ol 94%
3,4-Dimethyl-1-pentin-3-ol
Purity: 94%
CAS No.: 1482-15-1
Molecular Formula: (CH3)2CHC(OH)(CH3)C≡CH
Formula Weight: 112.17
Flash Point: 40°(104°F)
Boiling Point: 133°
UN No.: UN1987
MDL No.: MFCD00039845
Formel:
C₇H₁₂O Molecular Weight: 112.17 g/mol Siedepunkt: 133 °C Dichte: 0.87 Flammpunkt: 40 °C (104 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039845 CAS-Nummer: 1482-15-1 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...