[(4-Fluorophenyl)sulphonyl]acetonitrile 97%

Lieferant: Thermo Fisher Scientific
Gefahr

A19916.03 A19916.06
A19916.03EA 49.8 EUR
A19916.03 A19916.06
[(4-Fluorophenyl)sulphonyl]acetonitrile 97%
[(4-Fluorophenyl)sulphonyl]acetonitrile

Purity: 97% CAS No.: 32083-66-2 Molecular Formula: C8H6FNO2S Formula Weight: 199.20 Melting Point: 89-93° UN No.: UN3439 MDL No.: MFCD00179339

Formel: C₈H₆FNO₂S
Molecular Weight: 199.2 g/mol
Schmelzpunkt: 89…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00179339
CAS-Nummer: 32083-66-2
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR