5-Methyl-3-phenyl-1,2,4-oxadiazole 97%

Lieferant: Thermo Scientific

B20145.03 B20145.06
B20145.03EA 60.4 EUR
B20145.03 B20145.06
5-Methyl-3-phenyl-1,2,4-oxadiazole 97%
5-Methyl-3-phenyl-1,2,4-oxadiazole

Purity: 97% CAS No.: 1198-98-7 Molecular Formula: C9H8N2O Formula Weight: 223.28 Melting Point: 33-36° MDL No.: MFCD00085133

Formel: C₉H₈N₂O
Molecular Weight: 223,28 g/mol
Schmelzpunkt: 33…36 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00085133
CAS-Nummer: 1198-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR