1,3-Diphenyl-1H-pyrazol-5-amine 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Amino-1,3-diphenylpyrazole

B20464.06 B20464.03
B20464.06EA 48.5 EUR
B20464.06 B20464.03
1,3-Diphenyl-1H-pyrazol-5-amine 97%
1,3-Diphenyl-1H-pyrazol-5-amine

Purity: 97% CAS No.: 5356-71-8 Molecular Formula: C15H13N3 Formula Weight: 235.29 Odour: Odourless Melting Point: 129-132° MDL No.: MFCD00084878

Formel: C₁₅H₁₃N₃
Molecular Weight: 235,29 g/mol
Schmelzpunkt: 129…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084878
CAS-Nummer: 5356-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR