5-Chlor-2-thiophensulfonamid 97%

Lieferant: Thermo Scientific
Achtung

B20477.06EA 91.5 EUR
B20477.06 B20477.03
5-Chlor-2-thiophensulfonamid 97%
5-Chlor-2-thiophensulfonamid
Formel: C₄H₄ClNO₂S₂
Molecular Weight: 197,65 g/mol
Schmelzpunkt: 115…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052584
CAS-Nummer: 53595-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR