Ethyl-3,5-dimethylisoxazole-4-carboxylate 97%

Lieferant: Thermo Fisher Scientific

B20642.09 B20642.88
B20642.09EA 132 EUR
B20642.09 B20642.88
Ethyl-3,5-dimethylisoxazole-4-carboxylate 97%
Ethyl-3,5-dimethylisoxazole-4-carboxylate

Purity: 97% CAS No.: 17147-42-1 Molecular Formula: C8H11NO3 Formula Weight: 169.18 Boiling Point: 87-89°/2mm MDL No.: MFCD00052557

Formel: C₈H₁₁NO₃
Molecular Weight: 169.18 g/mol
Siedepunkt: 87…89 °C (2 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00052557
CAS-Nummer: 17147-42-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR